Silberchromat ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

40107.14
40107.14EA 284 EUR
40107.14
Silberchromat ≥99%
Silberchromat

Purity: 99% min CAS No.: 7784-01-2 Molecular Formula: Ag2CrO4 Formula Weight: 331.73 Physical Form: Powder Density: 5.63 Solubility: Slightly soluble in water. Soluble in HNO3 and ammonia Merck Reference: 14,8510 UN No.: 1479 MDL No.: MFCD00003402

Formel: Ag₂CrO₄
Siedepunkt: 1550 °C (1013 hPa)
Dichte: 5,63
Lagertemperatur: Raumtemperatur
MDL: MFCD00003402
CAS-Nummer: 7784-01-2
EINECS: 232-043-8
UN: 1479
ADR: 5.1,III
Merck Index: 14,8510

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR