Hypoxanthin, Millipore®
Lieferant: Merck Millipore Calbiochem
Synonyme:
1H-Purin-6-on, 6-Purinon, Purin-6(1H)-on, 6-Hydroxypurin
4010CBC-25GM
4010CBC-25GMEA
120
EUR
4010CBC-25GM
Hypoxanthin, Millipore®
Hypoxanthin
Formel:
C₅H₄N₄O Molecular Weight: 136,11 g/mol Schmelzpunkt: > 310 °C |
MDL:
MFCD00005725 CAS-Nummer: 68-94-0 EINECS: 200-697-3 Merck Index: 13,04895 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...