Silver hydrogen fluoride ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

40129.09
40129.09EA 121 EUR
40129.09
Silver hydrogen fluoride ≥99%
Silver hydrogen fluoride

Purity: 99% min CAS No.: 12249-52-4 Molecular Formula: AgF2H Formula Weight: 146.87 Physical Form: Crystalline Sensitivity: Light Sensitive; Hygroscopic Melting Point: dec. UN No.: UN1740 MDL No.: MFCD00042145

Formel: AgF₂H
Schmelzpunkt: Zersetzung
Lagertemperatur: Raumtemperatur
MDL: MFCD00042145
CAS-Nummer: 12249-52-4
UN: 1740
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR