Calciumiodid Hydrat
Lieferant: Thermo Fisher Scientific
|
Achtung
|
CAS No.: 71626-98-7
Molecular Formula: CaI2.xH2O
Formula Weight: 293.89(anhy)
Physical Form: Lump
Density: 2.55
Sensitivity: Hygroscopic
Appearance: White to grey
Melting Point: 42° dec.
Solubility: Freely soluble in water and alcohol
Merck Reference: 14,1677
MDL No.: MFCD00149622
Formel:
CaI₂·H₂O Dichte: 2,55 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149622 CAS-Nummer: 71626-98-7 EINECS: 233-276-8 Merck Index: 14,1677 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...