Calcium iodate monohydrate ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
40138.22
40138.36
40138.22EA
60.6
EUR
40138.22
40138.36
Calcium iodate monohydrate ≥99%
Calcium iodate monohydrate
Purity: 99+%
CAS No.: 10031-32-0
Molecular Formula: Ca(IO3)2.H2O
Formula Weight: 407.90 (389.88anhy)
Physical Form: Powder
Solubility: Slightly soluble in water
UN No.: UN1479
MDL No.: MFCD00036265
Formel:
Ca(IO₃)₂·H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036265 CAS-Nummer: 10031-32-0 EINECS: 232-191-3 UN: 1479 ADR: 5.1,II Merck Index: 14,1676 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...