Calciumsulfat, wasserfrei 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Calcium sulphate (anhydrous) anhydrite, Gypsum
40144.30
40144.A1
40144.A7
40144.30EA
50
EUR
40144.30
40144.A1
40144.A7
Calciumsulfat, wasserfrei 99%
Calcium sulphate
Purity: 99%
CAS No.: 7778-18-9
Molecular Formula: CaO4S
Formula Weight: 136.14
Physical Form: Powder
Density: 2.960
Appearance: White
Odour: Odourless
Melting Point: 1450°
Merck Reference: 14,1706
MDL No.: MFCD00010912
Drying agent, laboratory desiccant, paper filler
Formel:
CaO₄S Schmelzpunkt: 1450 °C Dichte: 2,960 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010912 CAS-Nummer: 7778-18-9 EINECS: 231-900-3 Merck Index: 14,1706 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...