Natriumdihydrogenphosphat ≥96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Natriumphosphat monobasisch
40145.30
40145.A1
40145.30EA
36.6
EUR
40145.30
40145.A1
Natriumdihydrogenphosphat ≥96%
Natriumdihydrogenphosphat
Purity: 96%
CAS No.: 7558-80-7
Molecular Formula: H2NaO4P
Formula Weight: 119.98
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Solubility: Freely soluble in water. Insoluble in alcohol
Merck Reference: 14,8660
MDL No.: MFCD00003527
Formel:
NaH₂PO₄ Molecular Weight: 119.98 g/mol Schmelzpunkt: ∼650 °C Dichte: 0.9996 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003527 CAS-Nummer: 7558-80-7 EINECS: 231-449-2 REACH: 01-2119489796-13 Merck Index: 12,08806 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...