Natriumdihydrogenphosphat ≥96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Natriumphosphat monobasisch

40145.30 40145.A1
40145.30EA 36.6 EUR
40145.30 40145.A1
Natriumdihydrogenphosphat ≥96%
Natriumdihydrogenphosphat

Purity: 96% CAS No.: 7558-80-7 Molecular Formula: H2NaO4P Formula Weight: 119.98 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Odour: Odourless Solubility: Freely soluble in water. Insoluble in alcohol Merck Reference: 14,8660 MDL No.: MFCD00003527

Formel: NaH₂PO₄
Molecular Weight: 119.98 g/mol
Schmelzpunkt: ∼650 °C
Dichte: 0.9996 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003527
CAS-Nummer: 7558-80-7
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR