N,O-Bis(trimethylsilyl)trifluoracetamid mit 10% Chlortrimethylsilan
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
40174.06
40174.14
40174.22
40174.06EA
48.8
EUR
40174.06
40174.14
40174.22
N,O-Bis(trimethylsilyl)trifluoracetamid mit 10% Chlortrimethylsilan
N,O-Bis(trimethylsilyl)trifluoracetamid mit 10% Chlortrimethylsilan
CAS No.: 25561-30-2
Molecular Formula: C8H18F3NOSi2
Formula Weight: 257.40
Storage Temperature: 2°C to 8°C
Flash Point: 23°(73°F)
Physical Form: Liquid
Sensitivity: Moisture Sensitive
Appearance: Colourless to pale yellow
Beilstein Reference: 3607620
UN No.: 2924
MDL No.: MFCD00008269
Formel:
C₈H₁₈F₃NOSi₂ Molecular Weight: 257.4 g/mol Flammpunkt: 23 °C (73 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00008269 CAS-Nummer: 25561-30-2 EINECS: 247-103-9 UN: 2924 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...