Kupfer(I)oxid ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99.9%
CAS No.: 1317-39-1
Molecular Formula: Cu2O
Formula Weight: 143.08
Physical Form: Powder
Density: 6.0
Refractive Index: 2.705
Sensitivity: Air and Moisture Sensitive
Appearance: Red to dark red
Odour: Odourless
Melting Point: 1235°
Boiling Point: 1800° dec.
Merck Reference: 14,2664
UN No.: 3077
MDL No.: MFCD00010974
Formel:
Cu₂O Siedepunkt: 1800 °C (Zers.) Schmelzpunkt: 1235 °C Dichte: 6,0 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010974 CAS-Nummer: 1317-39-1 EINECS: 215-270-7 UN: 3077 ADR: 9,III Merck Index: 14,2664 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...