Cadmium phosphate (ortho) ≥99%

Lieferant: Thermo Scientific
Gefahr

40201.18
40201.18EA 187 EUR
40201.18
Cadmium phosphate (ortho) ≥99%
Cadmium phosphate (ortho)

Purity: 99% CAS No.: 13847-17-1 Molecular Formula: Cd3(PO4)2 Formula Weight: 527.14 Physical Form: Powder Melting Point: ca 1500° Solubility: Insoluble in water UN No.: UN2570 MDL No.: MFCD00049414

Formel: Cd₃(PO₄)₂
Molecular Weight: 527,14 g/mol
Schmelzpunkt: ∼ 1500 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00049414
CAS-Nummer: 13847-17-1
EINECS: 237-581-7
UN: 2570
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR