Kupfer(II)wolframat ≥99,5% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

40217.18 40217.30
40217.18EA 103 EUR
40217.18 40217.30
Kupfer(II)wolframat ≥99,5% (Metall-Basis)
Kupfer(II)wolframat

Purity: 99.5% CAS No.: 13587-35-4 Molecular Formula: CuWO4 Formula Weight: 311.38 Physical Form: -200 Mesh Powder Odour: Odourless Merck Reference: 14,2657 MDL No.: MFCD00016067

Formel: CuO₄W
Lagertemperatur: Raumtemperatur
MDL: MFCD00016067
CAS-Nummer: 13587-35-4
EINECS: 237-022-7
Merck Index: 14,2657

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR