Germanium(IV)bromid ≥99,999% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr


Unter Argon in Ampullen verpackt

40219.09
40219.09EA 499 EUR
40219.09
Germanium(IV)bromid ≥99,999% (Metall-Basis)
Germanium(IV)bromid

Purity: 99.999% CAS No.: 13450-92-5 Molecular Formula: GeBr4 Formula Weight: 392.23 Physical Form: Low melting solid/Liquid, Ampouled under argon Density: 3.13 Appearance: White to gray Odour: Acidic Melting Point: 26° Boiling Point: 187° UN No.: 3260 MDL No.: MFCD00016115

Formel: Br₄Ge
Siedepunkt: 187 °C
Schmelzpunkt: 26 °C
Dichte: 3,13
Lagertemperatur: Raumtemperatur
MDL: MFCD00016115
CAS-Nummer: 13450-92-5
EINECS: 236-612-1
UN: 3260
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR