Cobaltselenid ≥99% (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr

Synonyme: Cobalt(II) selenide

40224.18
40224.18EA 223 EUR
40224.18
Cobaltselenid ≥99% (Metall-Basis)
Cobaltselenid

Purity: 99+% CAS No.: 1307-99-9 Molecular Formula: CoSe Formula Weight: 137.89 Physical Form: Powder Density: 7.650 Solubility: Soluble in HNO3, aqua regia UN No.: 3283 MDL No.: MFCD00016020

Formel: CoSe
Molecular Weight: 137,89 g/mol
Schmelzpunkt: 1055 °C
Dichte: 7,650
Lagertemperatur: Raumtemperatur
MDL: MFCD00016020
CAS-Nummer: 1307-99-9
EINECS: 215-155-1
UN: 3283
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR