Selenchlorid 99

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Selenium(I) chloride

ampuled under argon

40239.14 40239.6Y
40239.14EA 359 EUR
40239.14 40239.6Y
Selenchlorid 99
Selenchlorid

Purity: 99% CAS No.: 10025-68-0 Molecular Formula: Se2Cl2 Formula Weight: 228.83 Physical Form: Liquid, Ampouled under argon Density: 2.77 Melting Point: -85° Boiling Point: 127° dec. Solubility: Decomposes in water. Soluble in chloroform, benzene, carbon tetrachloride, carbon disulfide Merck Reference: 14,8432 UN No.: 2922 MDL No.: MFCD00011226

Formel: SeCl
Molecular Weight: 228.83 g/mol
Siedepunkt: 127 °C (Zers.)
Schmelzpunkt: –85 °C
Dichte: 2.77
Lagertemperatur: Raumtemperatur
MDL: MFCD00011226
CAS-Nummer: 10025-68-0
EINECS: 233-037-8
UN: 2922
ADR: 8 II
Merck Index: 14,8432

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR