Praseodym ≥99,5% (Metall-Basis ausgeschlossen Ta), Stäbe, Ø 12.7 mm (0.5 in)

Lieferant: Thermo Fisher Scientific

40296.DH 40296.DJ 40296.DM 40296.HF
40296.DHEA 616 EUR
40296.DH 40296.DJ 40296.DM 40296.HF
Praseodym ≥99,5% (Metall-Basis ausgeschlossen Ta), Stäbe, Ø 12.7 mm (0.5 in)
Praseodymium

Purity: 99.5% CAS No.: 7440-10-0 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.86g/mm, Cast rod, diameter is approximate Odour: Odourless

Formel: Pr
Lagertemperatur: Raumtemperatur
MDL: MFCD00011174
CAS-Nummer: 7440-10-0
EINECS: 231-120-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR