Praseodym ≥99,5% (Metall-Basis ausgeschlossen Ta), Stäbe, Ø 12.7 mm (0.5 in)
Lieferant: Thermo Fisher Scientific
40296.DH
40296.DJ
40296.DM
40296.HF
40296.DHEA
616
EUR
40296.DH
40296.DJ
40296.DM
40296.HF
Praseodym ≥99,5% (Metall-Basis ausgeschlossen Ta), Stäbe, Ø 12.7 mm (0.5 in)
Praseodymium
Purity: 99.5%
CAS No.: 7440-10-0
Molecular Formula: Packaged in mylar under argon
Physical Form: ≈0.86g/mm, Cast rod, diameter is approximate
Odour: Odourless
Formel:
Pr Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011174 CAS-Nummer: 7440-10-0 EINECS: 231-120-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...