Europium(III)trifluormethansulfonat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Europium(III) trifluoromethanesulphonate, Europium triflate
40305.09
40305.09EA
149
EUR
40305.09
Europium(III)trifluormethansulfonat ≥98%
Europium(III)trifluormethansulfonat
Purity: 98%
CAS No.: 52093-25-1
Molecular Formula: C3EuF9O9S3
Formula Weight: 599.15
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
MDL No.: MFCD00209587
Formel:
C₃EuF₉O₉S₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00209587 CAS-Nummer: 52093-25-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...