Holmium(III) trifluoromethanesulphonate hydrate ≥98%
Lieferant: Thermo Scientific
Achtung
40307.09
40307.09EA 183 EUR
40307.09
Holmium(III) trifluoromethanesulphonate hydrate ≥98%
Holmium(III) trifluoromethanesulphonate hydrate
Purity: 98% Molecular Formula: Ho(OSO2CF3)3.xH2O Formula Weight: 612.13(anhy) Physical Form: Powder MDL No.: MFCD00209590