Praseodym(III)trifluormethansulfonat 98
Lieferant: Thermo Fisher Scientific
|
Achtung
|
40310.09
40310.09EA
104
EUR
40310.09
Praseodym(III)trifluormethansulfonat 98
Praseodym(III)trifluormethansulfonat
Purity: 98%
CAS No.: 52093-27-3
Molecular Formula: Pr(OSO2CF3)3
Formula Weight: 588.11
Physical Form: Powder
Formel:
C₃F₉O₉PrS₃ Molecular Weight: 588.12 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00192417 CAS-Nummer: 52093-27-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...