Terbium(III)trifluormethansulfonat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
40312.04
40312.09
40312.04EA
88.3
EUR
40312.04
40312.09
Terbium(III)trifluormethansulfonat ≥98%
Terbium(III)trifluormethansulfonat
Purity: 98%
CAS No.: 148980-31-8
Molecular Formula: Tb(OSO2CF3)3
Formula Weight: 606.12
Physical Form: Powder
Formel:
C₃F₉O₉S₃Tb Lagertemperatur: Raumtemperatur |
MDL:
MFCD00192461 CAS-Nummer: 148980-31-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...