Yttrium(III)trifluormethansulfonat Hydrat
Lieferant: Thermo Fisher Scientific
|
Achtung
|
40315.04
40315.09
40315.04EA
35.6
EUR
40315.04
40315.09
Yttrium(III)trifluormethansulfonat Hydrat
Yttrium(III)trifluormethansulfonat Hydrat
CAS No.: 34629-25-9
Molecular Formula: Y(OSO2CF3)3.xH2O
Formula Weight: 536.10(anhy)
Physical Form: Powder
MDL No.: MFCD00209623
Formel:
C₃F₉O₉S₃Y·H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00209623 CAS-Nummer: 34629-25-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...