Cobalt(II)bromid, ultra trocken ≥99,99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Unter Argon in Ampullen verpackt
Purity: 99.99%
CAS No.: 7789-43-7
Molecular Formula: CoBr2
Formula Weight: 218.75
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 4.909
Appearance: Green
Melting Point: 678° (in N2)
Merck Reference: 14,2435
MDL No.: MFCD00016017
Formel:
Br₂Co Schmelzpunkt: 678 °C (in N{2}) Dichte: 4,909 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016017 CAS-Nummer: 7789-43-7 EINECS: 232-166-7 Merck Index: 14,2435 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...