Potassium fluorosulphate ≥99,5%

Lieferant: Thermo Fisher Scientific
Gefahr

40322.09 40322.18
40322.09EA 160 EUR
40322.09 40322.18
Potassium fluorosulphate ≥99,5%
Potassium fluorosulphate

Purity: 99.5% CAS No.: 13455-22-6 Molecular Formula: FKSO3 Formula Weight: 138.16 Physical Form: Powder/lump Sensitivity: Moisture Sensitive Appearance: White Melting Point: 350° dec. UN No.: UN3260 MDL No.: MFCD00216618

Formel: FKSO₃
Molecular Weight: 138.16 g/mol
Schmelzpunkt: 350 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00216618
CAS-Nummer: 13455-22-6
UN: 3260
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR