Ruthenium(IV)oxid ≥99,95% (Metall-Basis) Ru typ. 74%, Premion®, elektronische Qualität

Lieferant: Thermo Fisher Scientific
Gefahr

40336.02 40336.04 40336.09
40336.02EA 0 EUR
40336.02 40336.04 40336.09
Ruthenium(IV)oxid ≥99,95% (Metall-Basis) Ru typ. 74%, Premion®, elektronische Qualität
Ruthenium(IV)oxid

Purity: 99.95% CAS No.: 12036-10-1 Molecular Formula: RuO2 Formula Weight: 133.07 Physical Form: Sub-micron Powder, Surface Area 45-65m2/g Density: 6.97 Appearance: Black Odour: Odourless Melting Point: 1200° subl. UN No.: 1479 MDL No.: MFCD00011210


Semiconductor industry for the manufacture of resistor pastes

Formel: O₂Ru
Schmelzpunkt: 1200 °C (Subl.)
Dichte: 6,97
Lagertemperatur: Raumtemperatur
MDL: MFCD00011210
CAS-Nummer: 12036-10-1
EINECS: 234-840-6
UN: 1479
ADR: 5.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR