Dithalliumsulfat ≥99,5% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Thallium(I)sulfat

40342.18 40342.09 40342.30 40342.A1
40342.18EA 468 EUR
40342.18 40342.09 40342.30 40342.A1
Dithalliumsulfat ≥99,5% (Metall-Basis)
Dithalliumsulfat

Purity: 99.5% min CAS No.: 7446-18-6 Molecular Formula: Tl2SO4 Formula Weight: 504.80 Physical Form: Crystalline Density: 6.77 Melting Point: 632° Boiling Point: dec. Solubility: Solubility in water increases with temperature Merck Reference: 14,9269 UN No.: 1707 MDL No.: MFCD00011278

Formel: O₄STl₂
Molecular Weight: 504.83 g/mol
Siedepunkt: Zersetzung
Schmelzpunkt: 632
Dichte: 6.77
Lagertemperatur: Raumtemperatur
MDL: MFCD00011278
CAS-Nummer: 7446-18-6
EINECS: 231-201-3
UN: 1707
ADR: 6.1 II
Merck Index: 14,9269

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR