Strontiumchlorid Hexahydrat 99,0-103,0%, Kristalle, BAKER ANALYZED® ACS magnesiumarm, J.T.Baker®

Lieferant: Avantor
Achtung

4036-01EA 313.16 EUR
4036-01
Strontiumchlorid Hexahydrat 99,0-103,0%, Kristalle, BAKER ANALYZED® ACS magnesiumarm, J.T.Baker®
Strontiumchlorid Hexahydrat
Formel: SrCl₂·6H₂O
Siedepunkt: 100 °C (1013 hPa)
Schmelzpunkt: 61 °C
Dichte: 1,93 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149865
CAS-Nummer: 10025-70-4
EINECS: 233-971-6
Merck Index: 13,08920

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use 99.0 - 103.0 %
ACS - Assay (SrCl₂·6H₂O) 99.0 - 103.0 %
ACS - Insoluble Matter ≤0.005 %
ACS - pH of 5% Solution at 25°C 5.0 - 7.0
ACS - Sulfate (SO₄) ≤0.001 %
ACS - Barium (Ba) ≤0.05 %
ACS - Calcium (Ca) ≤0.05 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Trace Impurities - Magnesium (Mg) ≤2.000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR