Arsen(III)oxid ≥99,99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
40370.09
40370.18
40370.30
40370.09EA
71.7
EUR
40370.09
40370.18
40370.30
Arsen(III)oxid ≥99,99% (Metall-Basis)
Arsen(III)oxid
Purity: 99.99%
CAS No.: 1327-53-3
Molecular Formula: As2O3
Formula Weight: 197.84
Physical Form: -100 Mesh Powder
Density: 3.74
Melting Point: 312.3°
Boiling Point: 465° subl.
Merck Reference: 14,804
UN No.: 1561
MDL No.: MFCD00003433
Formel:
As₂O₃ Siedepunkt: 465 °C (Subl.) Schmelzpunkt: 312,3 °C Dichte: 3,74 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003433 CAS-Nummer: 1327-53-3 EINECS: 215-481-4 UN: 1561 ADR: 6.1,II Merck Index: 14,804 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...