Antimon(III)chlorid ≥99,999% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Gefahr

40372.04
40372.04EA 68.2 EUR
40372.04
Antimon(III)chlorid ≥99,999% (Metall-Basis), Puratronic®
Antimon(III)chlorid

Purity: 99.999% CAS No.: 10025-91-9 Molecular Formula: SbCl3 Formula Weight: 228.11 Physical Form: Crystalline Aggregates Density: 3.14 Melting Point: 73.4° Boiling Point: 223.5° Merck Reference: 14,707 UN No.: 1733 MDL No.: MFCD00011212

Formel: SbCl₃
Siedepunkt: 223,5 °C
Dichte: 3,14
Lagertemperatur: Raumtemperatur
MDL: MFCD00011212
CAS-Nummer: 10025-91-9
EINECS: 233-047-2
UN: 1733
ADR: 8,II
Merck Index: 14,707

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR