Bismut(III)chloridoxid ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Bismutoxychlorid
40396.18
40396.30
40396.A1
40396.18EA
52.3
EUR
40396.18
40396.30
40396.A1
Bismut(III)chloridoxid ≥98%
Bismuth chloride oxide
Purity: 98%
CAS No.: 7787-59-9
Molecular Formula: BiClO
Formula Weight: 260.43
Physical Form: Powder, typically 2 micron max
Density: 7.72
Refractive Index: 2.15
Appearance: Very pale brown
Odour: Odourless
Melting Point: dec.
Solubility: Practically insoluble in water, alcohol. Soluble in nitric acid, hydrochloric acid
Merck Reference: 14,1263
MDL No.: MFCD00010895
Used as pigment, in manufacture of artificial pearls, dry-cell cathodes
Formel:
BiClO Schmelzpunkt: Zersetzung Dichte: 7,72 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010895 CAS-Nummer: 7787-59-9 EINECS: 232-122-7 Merck Index: 14,1263 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...