Sodium tetracyanoplatinate(II) hydrate (≥48,4% Pt) ≥99,95% (Metall-Basis), Premion®
Lieferant: Thermo Scientific
|
Achtung
|
40400.03
40400.03EA
150
EUR
40400.03
Sodium tetracyanoplatinate(II) hydrate (≥48,4% Pt) ≥99,95% (Metall-Basis), Premion®
Sodium tetracyanoplatinate(II) hydrate
Purity: 99.95%
CAS No.: 699012-94-7
Molecular Formula: Na2Pt(CN)4.xH2O (x≈3)
Formula Weight: 345.14(anhy)
Physical Form: Crystalline powder
Appearance: White
Melting Point: Dec.
MDL No.: MFCD00798527
Formel:
Na₂Pt(CN)₄·xH₂O(x≈3) Molecular Weight: 345,14 g/mol (anhy) Schmelzpunkt: Zersetzung Lagertemperatur: Raumtemperatur |
MDL:
MFCD00798527 CAS-Nummer: 699012-94-7 EINECS: 239-536-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...