Platin(IV)iodid ≥99,95% (Metall-Basis) Pt ≥ 27.3%, Premion®
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Platintetraiodid
40401.06
40401.06EA
1023
EUR
40401.06
Platin(IV)iodid ≥99,95% (Metall-Basis) Pt ≥ 27.3%, Premion®
Platin(IV)iodid
Purity: 99.95%
CAS No.: 7790-46-7
Molecular Formula: I4Pt
Formula Weight: 702.70
Physical Form: Powder
Density: 6.06
Appearance: Grey to black
Melting Point: 130° dec.
Solubility: Decomposes in water. Soluble in alcohol, acetone, alkali, HI, KI, liquid NH3
UN No.: 3260
MDL No.: MFCD00016292
Formel:
I₄Pt Molecular Weight: 702,7 g/mol Schmelzpunkt: 130 °C (Zersetzung) Dichte: 6,06 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016292 CAS-Nummer: 7790-46-7 EINECS: 232-207-9 UN: 3260 ADR: 8,III Merck Index: 12,07685 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...