Iridium(IV)iodid ≥99,95% (Metall-Basis) Ir ≥ 27.0%, Premion®

Lieferant: Thermo Scientific
Achtung

40424.03
40424.03EA 220 EUR
40424.03
Iridium(IV)iodid ≥99,95% (Metall-Basis) Ir ≥ 27.0%, Premion®
Iridium(IV)iodid

Purity: 99.95% CAS No.: 7790-45-6 Molecular Formula: IrI4 Formula Weight: 699.82 Physical Form: Powder Solubility: Insoluble in water and alcohol. Soluble in aqueous KI MDL No.: MFCD00049960

Formel: I₄Ir
Molecular Weight: 699,84 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00049960
CAS-Nummer: 7790-45-6
EINECS: 232-206-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR