Sodium tetrabromoaurate(III) hydrate ≥99,99% (Metall-Basis), Premion®
Lieferant: Thermo Scientific
|
Achtung
|
40428.03
40428.03EA
128
EUR
40428.03
Sodium tetrabromoaurate(III) hydrate ≥99,99% (Metall-Basis), Premion®
Sodium tetrabromoaurate(III) hydrate
Purity: 99.99%
CAS No.: 10378-49-1
Molecular Formula: NaAuBr4.xH2O (x~2)
Formula Weight: 539.59(anhy)
Physical Form: Crystalline
Odour: Odourless
Solubility: Soluble in water
MDL No.: MFCD00274624
Formel:
NaAuBr₄·xH₂O(x~2) Molecular Weight: 539,57 g/mol (anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00274624 CAS-Nummer: 10378-49-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...