Glycerin, wasserfrei ≥99,5%, ultrareines Bioreagenz für die Biotechnologie, für die Elektrophorese, J.T.Baker®
Lieferant: Avantor
Synonyme:
1,2,3-Propantriol, Glycerol
4043-00EA
100.92
EUR
4043-00
4043-01
Glycerin, wasserfrei ≥99,5%, ultrareines Bioreagenz für die Biotechnologie, für die Elektrophorese, J.T.Baker®
Glycerin
Glycerol, anhydrous, Ultrapure Bioreagent, for enzyme stabilisation, freezing and electrophoresis buffers.
ULTRAPURE BIOREAGENT—A line of high purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography. Where applicable, tests include DNase, RNase, Protease, heavy metals and insoluble matter.
Formel:
HOCH₂CH(OH)CH₂OH Molecular Weight: 92.09 g/mol Siedepunkt: 290 °C (1013 hPa) Schmelzpunkt: 18.6 °C Dichte: 1.26 g/cm³ (20 °C) Flammpunkt: 177 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004722 CAS-Nummer: 56-81-5 EINECS: 200-289-5 Merck Index: 13,04497 |
|
Spezifikation Testergebnisse
For Laboratory,Research,or Manufacturing Use |
|
Assay (HOCH₂CHOHCH₂OH) | ≥99.5 % |
Absorbance of a 1M Solution - 280 nm | ≤0.050 |
DNase Activity (None Detected) | Passes Test |
RNase Activity (None Detected) | Passes Test |
Protease Activity (None Detected) | Passes Test |
Absorbance of a 1M Solution - Color (APHA) | ≤10 |
Residue after Ignition | ≤0.005 % |
Fatty Acid Esters (as Butyric Acid) | ≤0.05 % |
Trace Impurities - Iron (Fe) | ≤5 ppm |
Trace Impurities - Lead (Pb) | ≤2.000 ppm |
Trace Impurities - Mercury (Hg) | ≤10.000 ppm |
Trace Impurities - Zinc (Zn) | ≤2.000 ppm |
Vegetable Based |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...