Wasserstofftetranitrataurat(III)-hydrat (38,9 - 40,1% Au) ≥99,99% (Metall-Basis), Premion®

Lieferant: Thermo Scientific
Gefahr

40431.06
40431.06EA 798 EUR
40431.06
Wasserstofftetranitrataurat(III)-hydrat (38,9 - 40,1% Au) ≥99,99% (Metall-Basis), Premion®
Hydrogen tetranitratoaurate(III) hydrate

Decomposes in water. Soluble in HNO₃


Purity: 99.99% Molecular Formula: HAu(NO3)4.xH2O (x≈3) Formula Weight: 445.99(anhy) Physical Form: Crystalline Melting Point: ca 72° dec. Solubility: Decomposes in water. Soluble in HNO3 UN No.: UN1477 MDL No.: MFCD00046178

Formel: HAu(NO₃)₄·xH₂O(x≈3)
Molecular Weight: 445,99 g/mol (anhy)
Schmelzpunkt: ∼ 72 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00046178
UN: 1477
ADR: 5.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR