Gold(I)chlorid ≥99,99% (Metall-Basis) Au ≥ 84.2%, Premion®
Lieferant: Thermo Fisher Scientific
40432.01
40432.03
40432.06
40432.01EA
94.7
EUR
40432.01
40432.03
40432.06
Gold(I)chlorid ≥99,99% (Metall-Basis) Au ≥ 84.2%, Premion®
Gold(I)chlorid
Purity: 99.99%
CAS No.: 10294-29-8
Molecular Formula: AuCl
Formula Weight: 232.42
Physical Form: Powder
Density: 7.57
Melting Point: 289° dec.
Merck Reference: 14,4515
UN No.: 3260
MDL No.: MFCD00046175
Formel:
AuCl Dichte: 7,57 |
MDL:
MFCD00046175 CAS-Nummer: 10294-29-8 EINECS: 233-655-8 UN: 3260 ADR: 8,III Merck Index: 14,4515 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...