Tris(dimethylamino)gallium(III) dimer ≥99,9% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
Unter Argon in Ampullen verpackt
40443.27
40443.27EA
445
EUR
40443.27
Tris(dimethylamino)gallium(III) dimer ≥99,9% (Metall-Basis)
Tris(dimethylamino)gallium(III) dimer
Purity: 99.9%
CAS No.: 57731-40-5
Molecular Formula: C12H36Ga2N6
Formula Weight: 403.90
Physical Form: Powder/chunks, Ampouled under argon
Sensitivity: Moisture Sensitive
Appearance: White to yellow
Melting Point: 91°
Boiling Point: 125°/0.01mm subl.
UN No.: UN3131
MDL No.: MFCD00798535
Applications: Gallium nitride film precursor
Formel:
C₁₂H₃₆Ga₂N₆ Molecular Weight: 403,90 g/mol Siedepunkt: 125 °C (0,01 mmHg) Schmelzpunkt: 91 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00798535 CAS-Nummer: 57731-40-5 UN: 3131 ADR: 4.3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...