1-(4-Nitrophenylazo)-2-naphthol, Supelco®

Lieferant: Merck

Synonyme: Para Red, 1-((4-Nitrophenyl)diazenyl)naphthalen-2-ol

M-Clarity™ Qualitätsstufe = MQ100

40446-100MG
40446-100MGEA 265 EUR
40446-100MG
1-(4-Nitrophenylazo)-2-naphthol, Supelco®
1-(4-Nitrophenylazo)-2-naphthol
Formel: C₁₆H₁₁N₃O₃
Molecular Weight: 293.28 g/mol
MDL: MFCD00003908
CAS-Nummer: 6410-10-2
EINECS: 229-093-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76efc2a244d220ccd9a1d089770dff6d9e53a356&vid=ba10ca7d6e931ac79bbd409736487d203ecf0e62&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR