Zirkonium(IV)oxid ≥99,4% Metall-Basis ausgeschlossen Hf stabilisiert, Pulver -100+325 Mesh

Lieferant: Thermo Fisher Scientific
Achtung


Stabilisierung: Stabilisiert mit Calciumoxid (CaO) 4%

40453.18 40453.A1
40453.18EA 42.2 EUR
40453.18 40453.A1
Zirkonium(IV)oxid ≥99,4% Metall-Basis ausgeschlossen Hf stabilisiert, Pulver -100+325 Mesh
Zirkonium(IV)oxid

The main use of zirconia is in the production of ceramics. Zirconium dioxide is also used as the solid electrolyte in electrochromic devices As protective coating on particles of titanium dioxide pigments, as a refractory material, in insulation, abrasives and enamels.


Plasma Spray Grade. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, Alcohols, Halogens.

Formel: O₂Zr
Siedepunkt: ∼ 5000 °C
Schmelzpunkt: ∼ 2700 °C
Dichte: 5,75 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011310
CAS-Nummer: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR