8-Hydroxyjulolidine-9-carboxaldehyde 98%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: 9-Formyl-8-hydroxyjulolidine, 2,3,6,7-Tetrahydro-8-hydroxy-1H,5H-benzo[ij]quinolizine-9-carboxaldehyde, 8-Hydroxy-1,2,3,5,6,7-hexahydropyrido[3,2,1-ij]quinoline-9-carbaldehyde

M-Clarity™ Qualitätsstufe = MQ100

404640-1G
404640-1GEA 193 EUR
404640-1G
8-Hydroxyjulolidine-9-carboxaldehyde 98%, Sigma-Aldrich®
8-Hydroxyjulolidine-9-carboxaldehyde
Formel: C₁₃H₁₅NO₂
Molecular Weight: 217,27 g/mol
Schmelzpunkt: 72,..74 °C (Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00192477
CAS-Nummer: 63149-33-7
EINECS: 263-966-4

Bestellen

Spezifikation Testergebnisse

Appearance (Colour): Yellow to Tan to Green Conforms to Requirements
Appearance (Form) Powder or Crystals
Infrared spectrum Conforms to Structure
Carbon 70.0 - 73.7%
Nitrogen 6.2 - 6.6%
Purity (GC) ≥97.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR