Chromyl chloride ≥99,99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99.99%
CAS No.: 14977-61-8
Molecular Formula: CrO2Cl2
Formula Weight: 154.90
Physical Form: Liquid
Density: 1.911
Sensitivity: Moisture Sensitive
Odour: Pungent, makes eyes water
Melting Point: -96.5°
Boiling Point: 117°
UN No.: UN1758
MDL No.: MFCD00010951
Applications: Organic oxidations and chlorination, chromium coordination complexes, catalyst for polymerization of olefins
Formel:
CrO₂Cl₂ Siedepunkt: 117 °C Dichte: 1,911 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010951 CAS-Nummer: 14977-61-8 EINECS: 239-056-8 UN: 1758 ADR: 8,I Merck Index: 14,2248 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...