Bis(cyclopentadienyl)wolframdichlorid ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
unter Argon abgefüllt
40521.MD
40521.MDEA
92.5
EUR
40521.MD
Bis(cyclopentadienyl)wolframdichlorid ≥99%
Bis(cyclopentadienyl)wolframdichlorid
Purity: 99%
CAS No.: 12184-26-8
Molecular Formula: C10H10Cl2W
Formula Weight: 384.95
Physical Form: Powder, Packaged under argon
Sensitivity: Air and Moisture Sensitive
UN No.: 3261
MDL No.: MFCD00269852
Formel:
C₁₀H₁₀Cl₂W Molecular Weight: 384,93 g/mol Schmelzpunkt: >350 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00269852 CAS-Nummer: 12184-26-8 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...