Dysprosium(III)perchlorat Hexahydrat 50% (w/w) in wässriger Lösung, Reagenzqualität
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Dysprosium(III) perchlorate
40574.NA
40574.NAEA
56.4
EUR
40574.NA
Dysprosium(III)perchlorat Hexahydrat 50% (w/w) in wässriger Lösung, Reagenzqualität
Dysprosium(III)perchlorat Hexahydrat
CAS No.: 14692-17-2
Molecular Formula: Dy(ClO4)3.6H2O
Formula Weight: 568.94 (460.85anhy)
Physical Form: Liquid
Odour: Odourless
UN No.: 3211
MDL No.: MFCD00150227
Formel:
Cl₃DyO₁₂·6H₂O Molecular Weight: 568,94 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150227 CAS-Nummer: 14692-17-2 EINECS: 237-838-3 UN: 3211 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...