Holmium(III)perchlorat 50% (w/w) in wässriger Lösung, Reagenzqualität
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Holmium (III) perchlorate
40575.NC
40575.NCEA
264
EUR
40575.NC
Holmium(III)perchlorat 50% (w/w) in wässriger Lösung, Reagenzqualität
Holmium(III)perchlorat
CAS No.: 14017-54-0
Molecular Formula: Ho(ClO4)3.6H2O
Formula Weight: 571.37 (463.28anhy)
Physical Form: Liquid
Density: 1.57
Odour: Odourless
UN No.: 3211
MDL No.: MFCD00064643
Formel:
Cl₃HoO₁₂ Molecular Weight: 463,28 g/mol Dichte: 1,57 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064643 CAS-Nummer: 14017-54-0 EINECS: 237-839-9 UN: 3211 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...