Scandium(III)perchlorat 50% (w/w) in wässriger Lösung, Reagenzqualität
Lieferant: Thermo Scientific
|
Gefahr
|
40579.NA
40579.NAEA
107
EUR
40579.NA
Scandium(III)perchlorat 50% (w/w) in wässriger Lösung, Reagenzqualität
Scandium(III) perchlorate
CAS No.: 14066-05-8
Molecular Formula: Sc(ClO4)3.6H2O
Formula Weight: 451.42 (343.30anhy)
Physical Form: Liquid
Odour: Odourless
UN No.: 3211
MDL No.: MFCD00243289
Formel:
Cl₃O₁₂Sc Molecular Weight: 451,42 g/mol (343,30 anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00243289 CAS-Nummer: 14066-05-8 EINECS: 237-914-6 UN: 3211 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...