Antimon(III)kaliumoxidtartrat Trihydrat 99,0-103,0% ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
40619.22
40619.36
40619.14
40619.22EA
65.9
EUR
40619.22
40619.36
40619.14
Antimon(III)kaliumoxidtartrat Trihydrat 99,0-103,0% ACS
Antimon(III)kaliumoxidtartrat Trihydrat
Purity: 99.0-103.0%
CAS No.: 28300-74-5
Molecular Formula: C8H4K2O12Sb2.3H2O
Formula Weight: 667.87 (613.82anhy)
Physical Form: Powder
Density: 2.6
Appearance: White
Odour: Unpleasant
Merck Reference: 14,700
UN No.: 1551
MDL No.: MFCD00148863
Formel:
C₈H₄K₂O₁₂Sb₂·3H₂O Schmelzpunkt: 100 °C Dichte: 2,6 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00148863 CAS-Nummer: 28300-74-5 EINECS: 229-436-1 UN: 1551 ADR: 6.1,III Merck Index: 14,700 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...