Antimon(III)kaliumoxidtartrat Trihydrat 99,0-103,0% ACS

Lieferant: Thermo Fisher Scientific
Achtung

40619.22 40619.36 40619.14
40619.22EA 65.9 EUR
40619.22 40619.36 40619.14
Antimon(III)kaliumoxidtartrat Trihydrat 99,0-103,0% ACS
Antimon(III)kaliumoxidtartrat Trihydrat

Purity: 99.0-103.0% CAS No.: 28300-74-5 Molecular Formula: C8H4K2O12Sb2.3H2O Formula Weight: 667.87 (613.82anhy) Physical Form: Powder Density: 2.6 Appearance: White Odour: Unpleasant Merck Reference: 14,700 UN No.: 1551 MDL No.: MFCD00148863

Formel: C₈H₄K₂O₁₂Sb₂·3H₂O
Schmelzpunkt: 100 °C
Dichte: 2,6
Lagertemperatur: Raumtemperatur
MDL: MFCD00148863
CAS-Nummer: 28300-74-5
EINECS: 229-436-1
UN: 1551
ADR: 6.1,III
Merck Index: 14,700

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR