N-[(3-(Anilinomethylene)-2-chloro-1-cyclohexen-1-yl)methylene]aniline monohydrochloride 94%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: N-((2-Chloro-3-((phenylimino)methyl)cyclohex-2-en-1-ylidene)methyl)aniline hydrochloride

407232-5G
407232-5GEA 390 EUR
407232-5G
N-[(3-(Anilinomethylene)-2-chloro-1-cyclohexen-1-yl)methylene]aniline monohydrochloride 94%, Sigma-Aldrich®
N-[(3-(Anilinomethylene)-2-chloro-1-cyclohexen-1-yl)methylene]aniline monohydrochloride

N-[(3-(Anilinomethylene)-2-chloro-1-cyclohexen-1-yl)methylene]aniline monohydrochloride,94%

Formel: C₂H₁₉ClN₂
Molecular Weight: 359,29 g/mol
Schmelzpunkt: 224 °C (Zers., Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00191759
CAS-Nummer: 63857-00-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e3c127510b4c2810816326d59fb2cc94b33a7d3&vid=42a3e9772df98666839768673422244da730aee6&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR