Blei(II)oxid 99,999 Metals basis, Puratronic®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Bleimonoxid
40753.14
40753.22
40753.36
40753.14EA
86.9
EUR
40753.14
40753.22
40753.36
Blei(II)oxid 99,999 Metals basis, Puratronic®
Blei(II)oxid
Purity: 99.999%
CAS No.: 1317-36-8
Molecular Formula: PbO
Formula Weight: 223.19
Physical Form: Powder
Density: 9.53
Appearance: Yellow
Odour: Odourless
Melting Point: 886°
Merck Reference: 14,5413
UN No.: 3077
MDL No.: MFCD00011164
Formel:
PbO Molecular Weight: 223.2 g/mol Siedepunkt: 1470 Schmelzpunkt: 886 °C Dichte: 9.53 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011164 CAS-Nummer: 1317-36-8 EINECS: 215-267-0 UN: 3077 ADR: 9 III Merck Index: 14,5413 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...